missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11,11'-(Piperazine-1,4-diyl)bis(8-chloro-5H-dibenzo[b,e][1,4]-diazepine), Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
11,11'-(Piperazine-1,4-diyl)bis(8-chloro-5H-dibenzo[b,e][1,4]-diazepine)
Specifications
Specifications
| Chemical Name or Material | 11,11'-(Piperazine-1,4-diyl)bis(8-chloro-5H-dibenzo[b,e][1,4]-diazepine) |
| CAS | 263366-81-0 |
| Analyte or Component Names | 11,11'-(Piperazine-1,4-diyl)bis(8-chloro-5H-dibenzo[b,e][1,4]-diazepine) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H24 Cl2 N6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Clozapine Imp. B (EP), Clozapine Impurity B |
| InChI Formula | InChI=1S/C30H24Cl2N6/c31-19-9-11-25-27(17-19)35-29(21-5-1-3-7-23(21)33-25)37-13-15-38(16-14-37)30-22-6-2-4-8-24(22)34-26-12-10-20(32)18-28(26)36-30/h1-12,17-18,33-34H,13-16H2 |
| SMILES | Clc1ccc2Nc3ccccc3C(=Nc2c1)N4CCN(CC4)C5=Nc6cc(Cl)ccc6Nc7ccccc57 |
| IUPAC Name | 3-chloro-6-[4-(3-chloro-11H-benzo[b][1,4]benzodiazepin-6-yl)piperazin-1-yl]-11H-benzo[b][1,4]benzodiazepine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction