missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 10Z-Nonadecenoic acid

Product Code. 16454778
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454778 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16454778 Supplier MedChemExpress Supplier No. HY113450
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 10Z-Nonadecenoic acid is a kind of long-chain fatty acid with anti-tumor activity.

Specifications

Content And Storage Solution, -20°C, 2 years
Molecular Weight (g/mol) 296.49 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 73033-09-7
Product Type Natural Products
Purity 98.0%
Target Endogenous Metabolite
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.