missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Assay Buffer (10X)

Product Code. 10397784
Click to view available options
Quantity:
150 mL
Unit Size:
150mL
This item is not returnable. View return policy

Product Code. 10397784

Brand: Invitrogen™ T2187

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in immunoassays and in Western and Southern blotting for alkaline phosphatase Tropix™ substrate conjugates.

  • Quantity: 150mL
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Buffer
Quantity 150 mL
Form Liquid
Substrate Type Tropix
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.