Learn More
Leica Microsystems 10X/18 Eyepiece, DM100/300
Suitable eyepiece for the DM100/300 series of innovative educational compound microscopes. 10X/18 Eyepiece, DM100/300 is available with a reticule or pointer.
Brand: Leica Microsystems 13613333
Code : 04
Additional Details : Weight : 0.04500kg
Description
- For DM100/300 series of educational compound microscopes.
- Available with a reticule or pointer.
Order Info
MagnificationPower
Specifications
Eyepiece | |
DM100/300 Series Microscopes | |
10X |
18 | |
Reticule |