missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human SH3MD2 GST (N-Term) Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 57630 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 36.63 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | EC 6.3.2, EC 6.3.2.-, KIAA1494SH3 multiple domains protein 2, Plenty of SH3s, POSHFLJ21602, Protein POSH, putative E3 ubiquitin-protein ligase SH3RF1, RING finger protein 142, RNF142plenty of SH3 domains, SH3 domain containing ring finger 1, SH3 domain-containing RING finger protein 1, SH3 multiple domains 2, SH3MD2 |
| Gene Symbol | SH3RF1 |
| Research Category | Zinc Finger |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction