missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TREML1/TLT-1 (NP_835468) VersaClone cDNA

Product Code. 18704508
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18704508

Brand: R&D Systems™ RDC1884

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Triggering Receptor Expressed on Myeloid Cells-like 1

The Human TREML1/TLT-1 (NP_835468) VersaClone cDNA clone contains the complete ORF for TREML1/TLT-1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TREML1/TLT-1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifications

Product Type cDNA
Gene hTREML1
Accession Number NP_835468
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 949bp
Gene ID (Entrez) 340205 (Human); 71326 (Mouse)
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.