missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Sigma-1 R (NP_005857) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Human Sigma-1 R (NP_005857) VersaClone cDNA clone contains the complete ORF for Sigma-1 R/OPRS1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Sigma-1 R/OPRS1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
| Gene ID (Entrez) | 10280 |
| Gene Alias | Aging-associated gene 8 protein, ALS16, FLJ25585, hSigmaR1, OPRS1, SIG-1R, sigma 1, Sigma 1-type opioid receptor, sigma non-opioid intracellular receptor 1, sigma1 receptor, sigma1R, Sigma-1R, sigma1-receptor, SIGMAR1, SR31747 binding protein 1, SR31747-binding protein, SRBP, SR-BP, SR-BP1, SRBPMGC3851, type I sigma receptor |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction