missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ 10nm Streptavidin Gold Conjugate (10OD)

Product Code. p-7145987 Shop All Bio Techne Products
Click to view available options
Quantity:
0.2 mL
1 mL
Unit Size:
0.20mL
1mL
This item is not returnable. View return policy

Product Code. 18017523

Brand: Novus Biologicals™ 2520200

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Streptavidin gold nanoparticle conjugates can be used in conjunction with biotinylated reagents such as biotinylated antibodies or proteins, in applications such as lateral flow immunoassays or electron microscopy.

InnovaCoat GOLD nanoparticles have a proprietary surface coating which covalently binds antibodies, proteins, peptides or any other biomolecule with free amine groups to form highly stable conjugates. The surface coating can withstand the most extreme conditions (e.g. 2.5M NaOH at 70°C for 2 hours), making InnovaCoat™ GOLD an ideal reagent for a wide range of applications.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.2 mL
Content And Storage 4°C Do not freeze.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.