missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ 10nm Streptavidin Gold Conjugate (10OD)
Shop All Bio Techne Products
Click to view available options
Quantity:
0.2 mL
1 mL
Unit Size:
0.20mL
1mL
Description
InnovaCoat GOLD nanoparticles have a proprietary surface coating which covalently binds antibodies, proteins, peptides or any other biomolecule with free amine groups to form highly stable conjugates. The surface coating can withstand the most extreme conditions (e.g. 2.5M NaOH at 70°C for 2 hours), making InnovaCoat™ GOLD an ideal reagent for a wide range of applications.
Specifications
Specifications
| Quantity | 0.2 mL |
| Content And Storage | 4°C Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction