missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ 10nm Colloidal Gold Nanoparticles 10OD

Product Code. 18033464 Shop All Bio Techne Products
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
This item is not returnable. View return policy

Product Code. 18033464

Brand: Novus Biologicals™ 30100100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

10nm colloidal gold nanoparticles ultra high quality, with narrow size distribution and uniform shape

  • 10nm colloidal gold nanoparticles are ultra high quality, with narrow size distribution and uniform shape, supplied in range of sizes and concentrations for maximum flexibility
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 100 mL
Kit Contents One 20mL bottle at 10 OD
Product Type 10nm Colloidal Gold Nanoparticles 10OD Antibody Labeling Kit

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.