missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ 10nm Colloidal Gold Nanoparticles 10OD
Shop All Bio Techne Products
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
Description
- 10nm colloidal gold nanoparticles are ultra high quality, with narrow size distribution and uniform shape, supplied in range of sizes and concentrations for maximum flexibility
Specifications
Specifications
| Content And Storage | Store at 4°C. Do not freeze. |
| Quantity | 20 mL |
| Kit Contents | One 20mL bottle at 10 OD |
| Product Type | 10nm Colloidal Gold Nanoparticles 10OD Antibody Labeling Kit |
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction