missing translation for 'onlineSavingsMsg'
Learn More

Vinyltris(2-methoxyethoxy)silane, TRC

Product Code. 30449365
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 30449365

Brand: Toronto Research Chemicals TRCV75701310ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinyltris(2-methoxyethoxy)silane
CAS 1067-53-4
Molecular Formula C11 H24 O6 Si
Quantity 10 mL
Synonym Silane','tris(2-methoxyethoxy)vinyl- (6CI,7CI,8CI), 6-Ethenyl-6-(2-methoxyethoxy)-2,5,7,10-tetraoxa-6-silaundecane, A 172, A 172 (coupling agent), A 172NT, A 173, AC 77, CG 172, Crosile 172, DB 172, DB 172 (coupling agent), Dynasylan VTMOEO, FD 172, Fibnasil, GF 58, Geniosil GF 58, KBC 1003, KH 172, KH-A 172, NSC 78465, NUCA 172, SCA 972, SCA-A 172, SCA-V 71C, SG-SI 172, SH 6082, SZ 6082, Silane A 172, Silox VS 924, Silquest A 172, Silquest A 172NT, Struktol SCA 972, Tri(2-methoxyethoxy)vinylsilane, Tris(2-methoxyethoxy)vinylsilane, Tris(methoxyethoxy)vinylsilane, Tris(β-methoxyethoxy)vinylsilane, U 613, UNC-A 172, V 5000, VTMES-SCA 972, VTMOEO, VTS-ME, Vinyltri(methoxyethoxy)silane, Vinyltri(β-methoxyethoxy)silane, Vinyltris(2-methoxyethoxy)silane, Vinyltris(β-methoxyethoxy)silane, WD 27, WD 72, YGO 1204, Z 6082, Z 6172, 2,5,7,10-Tetraoxa-6-silaundecane','6-ethenyl-6-(2-methoxyethoxy)-
InChI Formula InChI=1S/C11H24O6Si/c1-5-18(15-9-6-12-2,16-10-7-13-3)17-11-8-14-4/h5H,1,6-11H2,2-4H3
SMILES COCCO[Si](OCCOC)(OCCOC)C=C
IUPAC Name ethenyl-tris(2-methoxyethoxy)silane
Molecular Weight (g/mol) 280.39
Formula Weight 280.1342
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.