missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 10 mL Nalgene Oak Ridge Tube or 12 mL Round Tube Adapters for Fiberlite F14-6x250 LE Rotor

Product Code. 11864511 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Fiberlite F14-6x250 LE rotor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11864511 Fiberlite F14-6x250 LE rotor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11864511 Supplier Thermo Scientific™ Supplier No. 00389
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Accommodate 10mL or 12mL tubes in the Thermo Scientific™ Fiberlite™ F14-6x250 LE rotor.

Accommodate 10mL Nalgene Oak Ridge or 12mL round tubes in the Thermo Scientific™ Fiberlite™ F14-6x250 LE rotor.

TRUSTED_SUSTAINABILITY

Specifications

Type Round Tube Adapter
Tube Type 10 mL Nalgene Oak Ridge Tube, 12 mL Round Tube
Capacity 10, 12 mL
For Use With (Equipment) Fiberlite F14-6x250 LE rotor
Description 10mL Nalgene Oak Ridge Tube or 12mL Round Tube Adapter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.