missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 10 mL Nalgene Oak Ridge Tube or 12mL Round Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
Description
Accommodate 10mL Nalgene Oak Ridge or 12mL round tubes in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.
Specifications
Specifications
| Type | Ridge Tube Adapter |
| Tube Type | 10 mL Nalgene Oak Ridge Tube, 12 mL Round Tube |
| Capacity | 10, 12 mL |
| For Use With (Equipment) | Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors |
| Description | 10mL Nalgene Oak Ridge Tube or 12mL Round Tube Adapter |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?