missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 10 mL Serum Transport Tubes
GREENER_CHOICE

Product Code. 17125301
Click to view available options
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 17125301

Brand: Fisherbrand™ M995200

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

10 mL threaded transport tube for liquid powders and product samples. They are fabricated from FDA-approved clarified polypropylene resin for visual clarity.

  • 10 mL screw top polypropylene tubes
  • Free-standing with skirted, round bottom
  • Graduation in 1 mL increments up to 8 mL
  • Non-sterile
  • 1000 per case


Note: Caps are not included; sold separately (See S40200, S40201, S40202, S40203, S40204)

TRUSTED_SUSTAINABILITY

Specifications

Description 10 mL transport tube, clear
Diameter (Metric) Outer 15 mm
Material Polypropylene
Sterility Non-sterile
Volume (Metric) 10 mL
No. per Case 1000
Style Skirted, round bottom
Closure Type Screw Cap
Height (Metric) 86 mm
Type Sample Tube
For Use With (Equipment) Capitol Vial Screw Caps (not included)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.