missing translation for 'onlineSavingsMsg'
Learn More

Zolmitriptan, TRC

Product Code. 30003928
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003928 10 mg 10mg
30137428 50 mg 50mg
30003331 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003928 Supplier Toronto Research Chemicals Supplier No. TRCZ63900010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139264-17-8
Molecular Formula C16 H21 N3 O2
Molecular Weight (g/mol) 287.36
Synonym Zolmitriptan, (4S)-4-[[3-[2-Dimethylamino)ethyl]-1H-indol-5-yl]methyl]oxazolidin-2-one, (S)-4-[[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]methyl]-2-oxazolidinone, 311C90, Asco Top, BW 311C90, Xolnox, Zipton, Zomig, Zominat
IUPAC Name (4S)-4-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1,3-oxazolidin-2-one
SMILES CN(C)CCc1c[nH]c2ccc(C[C@H]3COC(=O)N3)cc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H21N3O2/c1-19(2)6-5-12-9-17-15-4-3-11(8-14(12)15)7-13-10-21-16(20)18-13/h3-4,8-9,13,17H,5-7,10H2,1-2H3,(H,18,20)/t13-/m0/s1
Formula Weight 287.1634
Chemical Name or Material Zolmitriptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.