missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zeranol-d5 (Mixture of Diastereomers), TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Zeranol-d5 (Mixture of Diastereomers) |
| CAS | 1217532-71-2 |
| Molecular Formula | C18H21D5O5 |
| Quantity | 10 mg |
| Synonym | (3S)-3,4,5,6,7,8,9,10,11,12-Decahydro-7,14,16-trihydroxy-3-methyl -1H-2-benzoxacyclotetradecin-1-one-d5, Zearalanol-d5, Zeranol-d5, Ralabol-d5, Ralgro-d5, Ralone-d5 |
| InChI Formula | InChI=1S/C18H26O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h10-12,14,19-21H,2-9H2,1H3/t12-,14?/m0/s1/i8D2,9D2,14D |
| SMILES | O=C1C2=C(O)C=C(O)C=C2CCCCC([2H])([2H])C(O)([2H])C([2H])([2H])CC[C@H](C)O1 |
| IUPAC Name | (11S)-6,6,7,8,8-pentadeuterio-7,15,17-trihydroxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),15,17-trien-13-one |
| Molecular Weight (g/mol) | 327.43 |
| Formula Weight | 327.2094 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction