missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthophyll (~80%), TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Lutein |
| CAS | 127-40-2 |
| Molecular Formula | C40 H56 O2 |
| Quantity | 10 mg |
| Synonym | (3R,3'R,6'R)-beta,epsilon-Carotene-3,3'-diol, (3R,3'R,6'R)-Lutein, (all-E)-Lutein, 6'-Hydro-4',5'-dehydro-beta-carotene-3,3'-diol, Bo-Xan, E 161, E 161b, FloraGLO, FloraGLO Lutein, Lutein, Lutein A, Luteine, OS 24, Oro Glo 7, Vegetable lutein, Vegetable luteol, Xanthophyll, all-trans-(+)-Xanthophyll, all-trans-Lutein, all-trans-Xanthophyll, trans-Lutein |
| InChI Formula | InChI=1S/C40H56O2/c1-29(17-13-19-31(3)21-23-37-33(5)25-35(41)27-39(37,7)8)15-11-12-16-30(2)18-14-20-32(4)22-24-38-34(6)26-36(42)28-40(38,9)10/h11-25,35-37,41-42H,26-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+/t35-,36+,37-/m0/s1 |
| SMILES | C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C[C@@H](O)CC1(C)C)\C=C\C=C(/C)\C=C\[C@H]2C(=C[C@H](O)CC2(C)C)C |
| IUPAC Name | (1R)-4-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-[(1R,4R)-4-hydroxy-2,6,6-trimethylcyclohex-2-en-1-yl]-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaenyl]-3,5,5-trimethylcyclohex-3-en-1-ol |
| Molecular Weight (g/mol) | 568.87 |
| Formula Weight | 568.428 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction