missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-C59, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 4-(2-Methyl-4-pyridinyl)-N-[4-(3-pyridinyl)phenyl]-benzeneacetamide |
| CAS | 1243243-89-1 |
| Molecular Formula | C8 H11 N5 O3 |
| Quantity | 10 mg |
| Synonym | Wnt-C59, 4-(2-Methyl-4-pyridinyl)-N-[4-(3-pyridinyl)phenyl]-benzeneacetamide |
| InChI Formula | InChI=1S/C8H11N5O3/c1-5-6(13(15)16)7(14)11-8(10-5)9-4-12(2)3/h4H,1-3H3,(H,10,11,14)/b9-4- |
| SMILES | CN(C)\C=N/C1=NC(=C(C(=O)N1)[N+](=O)[O-])C |
| IUPAC Name | N,N-dimethyl-N'-(4-methyl-5-nitro-6-oxo-1H-pyrimidin-2-yl)methanimidamide |
| Molecular Weight (g/mol) | 225.205 |
| Formula Weight | 225.086 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction