missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vonoprazan Fumarate, TRC
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Specifications
Specifications
| Chemical Name or Material | Vonoprazan Fumarate |
| CAS | 881681-01-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H16 F N3 O2 S . C4 H4 O4 |
| Quantity | 10 mg |
| Synonym | 1H-Pyrrole-3-methanamine','5-(2-fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-','(2E)-2-butenedioate (1:1), 5-(2-Fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-1H-pyrrole-3-methanamine (2E)-2-butenedioate (1:1), TAK 438, TAK 438 monofumarate, Takecab, Vocinti, Vonoprazan fumarate, 5-(2-Fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-1H-pyrrole-3-methanamine fumarate, 1-[5-(2-Fluorophenyl)-1-[(pyridin-3-yl)sulfonyl]-1H-pyrrol-3-yl]-N-methylmethanamine fumarate, 1H-Pyrrole-3-methanamine','5-(2-fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-','(2E)-2-butenedioate (1:1), 5-(2-Fluorophenyl)-N-methyl-1-(3-pyridinylsulfonyl)-1H-pyrrole-3-methanamine (2E)-2-butenedioate (1:1), TAK 438, TAK 438 monofumarate, Takecab, Voci |
| InChI Formula | InChI=1S/C17H16FN3O2S.C4H4O4/c1-19-10-13-9-17(15-6-2-3-7-16(15)18)21(12-13)24(22,23)14-5-4-8-20-11-14;5-3(6)1-2-4(7)8/h2-9,11-12,19H,10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+ |
| SMILES | CNCc1cc(c2ccccc2F)n(c1)S(=O)(=O)c3cccnc3.OC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-but-2-enedioic acid;1-[5-(2-fluorophenyl)-1-pyridin-3-ylsulfonylpyrrol-3-yl]-N-methylmethanamine |
| Molecular Weight (g/mol) | 461.463 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur