missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VLX600, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 1-(2-Pyridinyl)ethanone 2-(6-Methyl-5H-1,2,4-triazino[5,6-b]indol-3-yl)hydrazone |
| CAS | 327031-55-0 |
| Molecular Formula | C17H15N7 |
| Quantity | 10 mg |
| Synonym | 1-(2-Pyridinyl)ethanone 2-(6-Methyl-5H-1,2,4-triazino[5,6-b]indol-3-yl)hydrazone, VLX600 |
| InChI Formula | InChI=1S/C17H15N7/c1-10-6-5-7-12-14(10)19-16-15(12)22-24-17(20-16)23-21-11(2)13-8-3-4-9-18-13/h3-9H,1-2H3,(H2,19,20,23,24)/b21-11+ |
| SMILES | C\C(=N/Nc1nnc2c3cccc(C)c3[nH]c2n1)\c4ccccn4 |
| IUPAC Name | 6-methyl-N-[(E)-1-pyridin-2-ylethylideneamino]-5H-[1,2,4]triazino[5,6-b]indol-3-amine |
| Molecular Weight (g/mol) | 317.35 |
| Formula Weight | 317.1389 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction