missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin D2-d3, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Ergocalciferol-(6,19,19-d3) |
| CAS | 1217448-46-8 |
| Molecular Formula | C28 2H3 H41 O |
| Quantity | 10 mg |
| Synonym | D3-Vitamin D2 (6,19,19-D3) |
| InChI Formula | InChI=1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1/i4D2,13D |
| SMILES | [2H]C(=C1CC[C@H](O)C/C/1=C(\[2H])/C=C/2\CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)\C=C\[C@H](C)C(C)C)[2H] |
| IUPAC Name | (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]-1-deuterioethylidene]-4-(dideuteriomethylidene)cyclohexan-1-ol |
| Molecular Weight (g/mol) | 399.67 |
| Formula Weight | 399.358 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction