missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin A-d5 Acetate, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Vitamin A-d5 Acetate |
| CAS | 1217219-62-9 |
| Molecular Formula | C22H27D5O2 |
| Quantity | 10 mg |
| Synonym | (all-E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl-d5)-2-4,6,8-nonatetraen- 1-ol Acetate, Retinyl-d5 Acetate, Vitamin A1-d5 Acetate, all-trans-Retinol-d5 Acetate |
| InChI Formula | InChI=1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/i3D3,11D2 |
| SMILES | [2H]C1([2H])CCC(C)(C)C(/C=C/C(C)=C/C=C/C(C)=C/COC(C)=O)=C1C([2H])([2H])[2H] |
| IUPAC Name | [9-[3,3-dideuterio-6,6-dimethyl-2-(trideuteriomethyl)cyclohexen-1-yl]-3,7-dimethylnona-2,4,6,8-tetraenyl] acetate |
| Molecular Weight (g/mol) | 333.52 |
| Formula Weight | 333.2716 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction