missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Varenicline N-Glucoside, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Varenicline N-Glucoside |
| CAS | 873302-31-9 |
| Molecular Formula | C19 H23 N3 O5 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C19H23N3O5/c23-8-15-16(24)17(25)18(26)19(27-15)22-6-9-3-10(7-22)12-5-14-13(4-11(9)12)20-1-2-21-14/h1-2,4-5,9-10,15-19,23-26H,3,6-8H2/t9?,10?,15-,16-,17+,18-,19?/m1/s1 |
| SMILES | OC[C@H]1OC([C@H](O)[C@@H](O)[C@@H]1O)N2CC3CC(C2)c4cc5nccnc5cc34 |
| Molecular Weight (g/mol) | 373.4 |
| Formula Weight | 373.1638 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction