missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vardenafil-d5, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Vardenafil-D5 |
| CAS | 1189685-70-8 |
| Molecular Formula | C23 2H5 H27 N6 O4 S |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C23H32N6O4S/c1-5-8-20-24-16(4)21-23(30)25-22(26-29(20)21)18-15-17(9-10-19(18)33-7-3)34(31,32)28-13-11-27(6-2)12-14-28/h9-10,15H,5-8,11-14H2,1-4H3,(H,25,26,30)/i2D3,6D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])N1CCN(CC1)S(=O)(=O)c2ccc(OCC)c(c2)C3=Nn4c(CCC)nc(C)c4C(=O)N3 |
| IUPAC Name | 2-[2-ethoxy-5-[4-(1,1,2,2,2-pentadeuterioethyl)piperazin-1-yl]sulfonylphenyl]-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one |
| Molecular Weight (g/mol) | 493.63 |
| Formula Weight | 493.252 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction