missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valproic Acid-d4, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Valproic acid 4,4,4',4'-d4 |
| CAS | 87745-17-3 |
| Molecular Formula | C8 D4 H12 O2 |
| Quantity | 10 mg |
| Synonym | Pentanoic-3,3-d2 acid','2-(propyl-1,1-d2)- (9CI), 3,3,3',3'-Tetradeuterovalproic acid, 2-(Propyl-1,1-d2)pentanoic-3,3-d2 acid, Valproic acid-d4, Valproic-3,3,3',3'-D4 acid |
| InChI Formula | InChI=1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)/i3D2,4D2 |
| SMILES | [2H]C([2H])(C)CC(CC([2H])([2H])C)C(=O)O |
| IUPAC Name | 4,4-dideuterio-2-(2,2-dideuteriopropyl)pentanoic acid |
| Molecular Weight (g/mol) | 148.24 |
| Formula Weight | 148.1401 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction