missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trospium-d8 Chloride, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Trospium-d8 Chloride |
| Molecular Formula | C25H22D8ClNO3 |
| Quantity | 10 mg |
| Synonym | (1α,3β,5α)-3-[(2-Hydroxy-2,2-diphenylacetyl)oxy]-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-pyrrolidinium]-d8 Chloride, Azoniaspiro-d8, Keptan-d8, Relaspium-d8, Sanctura-d8, Spasmex-d8, Spasmo 3-d8 |
| InChI Formula | InChI=1S/C25H30NO3.ClH/c27-24(25(28,19-9-3-1-4-10-19)20-11-5-2-6-12-20)29-23-17-21-13-14-22(18-23)26(21)15-7-8-16-26;/h1-6,9-12,21-23,28H,7-8,13-18H2;1H/q+1;/p-1/t21-,22+,23?;/i7D2,8D2,15D2,16D2; |
| SMILES | O=C(C(O)(C1=CC=CC=C1)C2=CC=CC=C2)O[C@H]3C[C@H]4CC[C@@H](C3)[N+]45C([2H])([2H])C([2H])([2H])C([2H])([2H])C5([2H])[2H].[Cl-] |
| Molecular Weight (g/mol) | 436.01 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction