missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triptolide-d3 (major) (>90%), TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Triptolide-d3 |
| Molecular Formula | C20H21D3O6 |
| Quantity | 10 mg |
| Synonym | (3bS,4aS,5aS,6R,6aR,7aS,7bS,8aS,8bS)-3b,4,4a,6,6a,7a,7b,8b,9,10-Decahydro -6-hydroxy-8b-methyl-6a-(1-methylethyl)-trisoxireno[4b,5:6,7:8a,9]phenanthro[1,2-c]furan-1(3H)-one-d3, (-)-Triptolide-d3, Triptolid-d3 |
| SMILES | O[C@H]1[C@]2(O3)[C@@H]3C[C@@]4([2H])C5=C(C(OC5([2H])[2H])=O)CC[C@]4(C)[C@]2(O6)[C@@H]6[C@H]7C1(O7)C(C)C |
| Molecular Weight (g/mol) | 363.42 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction