missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimipramine-d3 (Major) Maleate Salt, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Trimipramine-D3 maleate |
| CAS | 1185245-93-5 |
| Molecular Formula | C20 2H3 H23 N2 . C4 H4 O4 |
| Quantity | 10 mg |
| Synonym | Trimipramine-D3 Maleate |
| InChI Formula | InChI=1S/C20H26N2.C4H4O4/c1-16(14-21(2)3)15-22-19-10-6-4-8-17(19)12-13-18-9-5-7-11-20(18)22;5-3(6)1-2-4(7)8/h4-11,16H,12-15H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/i2D3; |
| SMILES | [2H]C([2H])([2H])N(C)CC(C)CN1c2ccccc2CCc3ccccc13.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,2-dimethyl-N-(trideuteriomethyl)propan-1-amine |
| Molecular Weight (g/mol) | 413.52 |
| Formula Weight | 413.2394 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction