missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimeprazine-d6 Hemitartrate Salt, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Trimeprazine-d6 Hemitartrate Salt |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 2H6 H16 N2 S |
| Quantity | 10 mg |
| Synonym | 2-Methyl-3-phenothiazin-10-yl-N,N-bis(trideuteriomethyl)propan-1-amine |
| InChI Formula | InChI=1S/C18H22N2S/c1-14(12-19(2)3)13-20-15-8-4-6-10-17(15)21-18-11-7-5-9-16(18)20/h4-11,14H,12-13H2,1-3H3/i2D3,3D3 |
| SMILES | [2H]C([2H])([2H])N(CC(C)CN1c2ccccc2Sc3ccccc13)C([2H])([2H])[2H] |
| IUPAC Name | 2-methyl-3-phenothiazin-10-yl-N,N-bis(trideuteriomethyl)propan-1-amine |
| Molecular Weight (g/mol) | 304.48 |
| Formula Weight | 304.188 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction