missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triglyme-d6, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Triglyme-d6 |
| CAS | 1219192-16-1 |
| Molecular Formula | C8H12D6O4 |
| Quantity | 10 mg |
| Synonym | 2,5,8,11-Tetraoxadodecane-d6, 1,2-Bis(2-methoxyethoxy)ethane-d6, Ansul Ether 161-d6, DMTG-d6, Glyme 4, Hisolve MTM-d6, Methyltriglyme-d6, NSC 66400-d6, Triethylene Glycol Dimethyl-d6 Ether |
| InChI Formula | InChI=1S/C8H18O4/c1-9-3-5-11-7-8-12-6-4-10-2/h3-8H2,1-2H3/i1D3,2D3 |
| SMILES | [2H]C([2H])([2H])OCCOCCOCCOC([2H])([2H])[2H] |
| IUPAC Name | 2,5,8,11-tetraoxadodecane-1,1,1,12,12,12-d6 |
| Molecular Weight (g/mol) | 184.26 |
| Formula Weight | 184.16 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction