missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylenephosphoramide-d12, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Triethylenephosphoramide-d12 |
| CAS | 1246816-29-4 |
| Molecular Formula | C6D12N3OP |
| Quantity | 10 mg |
| Synonym | 1,1',1''-Phosphinylidynetrisaziridine-d12, Tris(1-aziridinyl)phosphine-d12 Oxide, APO-d12, Aphoxide-d12, ENT 24915-d12, NSC 9717-d12, Phosphoric Acid Triethylene Imide-d12, N,N',N''-Tri-1,2-ethanediylphosphoric Triamide-d12, TAPO-d12, TEF-d12, TEPA-d12, Tri-1-aziridinyl-phosphine-d12 Oxide, Triaziridinophosphine-d12 Oxide, Triaziridinylphosphine-d12 Oxide, Triethylenephosphorotriamide-d12, Tris(1-aziridine)phosphine-d12 Oxide |
| InChI Formula | InChI=1S/C6H12N3OP/c10-11(7-1-2-7,8-3-4-8)9-5-6-9/h1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2 |
| SMILES | O=P(N1C([2H])([2H])C1([2H])[2H])(N2C([2H])([2H])C2([2H])[2H])N3C([2H])([2H])C3([2H])[2H] |
| IUPAC Name | 1-bis(2,2,3,3-tetradeuterioaziridin-1-yl)phosphoryl-2,2,3,3-tetradeuterioaziridine |
| Molecular Weight (g/mol) | 185.23 |
| Formula Weight | 185.1471 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction