missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triazophos-D5, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Triazophos D5 (Phenyl D5) |
| CAS | 1773496-62-0 |
| Molecular Formula | C12 2H5 H11 N3 O3 P S |
| Quantity | 10 mg |
| Synonym | Triazophos D5 (Phenyl D5) |
| InChI Formula | InChI=1S/C12H16N3O3PS/c1-3-16-19(20,17-4-2)18-12-13-10-15(14-12)11-8-6-5-7-9-11/h5-10H,3-4H2,1-2H3/i5D,6D,7D,8D,9D |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])n2cnc(OP(=S)(OCC)OCC)n2 |
| IUPAC Name | diethoxy-[[1-(2,3,4,5,6-pentadeuteriophenyl)-1,2,4-triazol-3-yl]oxy]-sulfanylidene-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 318.34 |
| Formula Weight | 318.0964 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction