missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tauro-d4-ursodeoxycholic Acid, TRC
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Specifications
Specifications
| Chemical Name or Material | Tauro-d4-ursodeoxycholic Acid |
| Molecular Formula | C26H41D4NO6S |
| Quantity | 10 mg |
| Synonym | 2-[[(3α,5β,7β)-3,7-Dihydroxy-24-oxocholan-24-yl]amino]ethane-1,1,2,2-d4-sulfonic acid |
| InChI Formula | InChI=1S/C26H45NO6S/c1-16(4-7-23(30)27-12-13-34(31,32)33)19-5-6-20-24-21(9-11-26(19,20)3)25(2)10-8-18(28)14-17(25)15-22(24)29/h16-22,24,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33)/t16-,17+,18-,19-,20+,21+,22+,24+,25+,26-/m1/s1/i12D2,13D2 |
| SMILES | C[C@H](CCC(NC([2H])(C([2H])(S(=O)(O)=O)[2H])[2H])=O)[C@@]1([H])CC[C@]2([H])[C@@]1(CC[C@@]3([H])[C@@]2([H])[C@H](C[C@@]4([H])[C@@]3(CC[C@H](C4)O)C)O)C |
| IUPAC Name | 2-((R)-4-((3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulfonic-1,1,2,2-d4 acid |
| Molecular Weight (g/mol) | 503.73 |
| Formula Weight | 503.32 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction