missing translation for 'onlineSavingsMsg'
Learn More

Tacedinaline, MedChemExpress

Product Code. 16458148
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16458148

Brand: Medchem Express HY5093410mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tacedinaline (N-acetyldinaline) is an inhibitor of the histone deacetylase (HDAC) with IC50s of 0.9, 0.9, 1.2 μM for recombinant HDAC 1, 2 and 3 respectively.

Tacedinaline (N-acetyldinaline) is an inhibitor of the histone deacetylase (HDAC) with IC50s of 0.9, 0.9, 1.2 μM for recombinant HDAC 1, 2 and 3 respectively.

Specifications

Chemical Name or Material Tacedinaline
Physical Form Solid
CAS 112522-64-2
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H15N3O2
For Use With (Application) Cancer-programmed cell death
Synonym N-acetyldinaline CI-994 Goe-5549
Solubility Information DMSO : ≥ 58 mg/mL (215.37 mM)
SMILES O=C(NC1=C(N)C=CC=C1)C2=CC=C(NC(C)=O)C=C2
Molecular Weight (g/mol) 269.3
Formula Weight 269.3
Percent Purity 95.77%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.