missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TAC
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TAC is a telomerase reverse transcriptase (TERT) activator. TAC upregulates TERT gene transcription via the MEK/ERK/AP-1 cascade. In primary human cells and naturally aged mice, TAC enhances telomere synthesis, reduces tissue aging markers, cellular senescence, and inflammatory cytokines. In the brain, TAC induces neurogenesis, reduces neuroinflammation, and preserves cognitive function.
Specifications
Tekniske data
| Chemical Name or Material | N-(3-Chloro-4-fluorophenyl)-4-fluoro-3,5-dimethylbenzenesulfonamide |
| CAS | 666699-46-3 |
| Quantity | 10 mg |
| Target | TERT |
| Molecular Formula | C14H12ClF2NO2S |
| Purity | 0.98 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion