missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salvinorin A, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Salvinorin A
Specifications
Specifications
| Chemical Name or Material | Salvinorin A |
| CAS | 83729-01-5 |
| Analyte or Component Names | Salvinorin A |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H28 O8 |
| Synonym | 2 H-Naphtho[2,1-c]pyran-7-carboxylic acid, 9-(acetyloxy)-2-(3-furanyl)dodecahydro-6 A,10 b-dimethyl-4,10-dioxo-, methyl ester, (2 S,4 aR,6 aR,7 R,9 S,10 AS,10 bR)-, 2 H-Naphtho[2,1-c]pyran-7-carboxylic acid, 9-(acetyloxy)-2-(3-furanyl)dodecahydro-6 A,10 b-dimethyl-4,10-dioxo-, methyl ester, [2 S-(2α,4 Aα,6 Aβ,7β,9β,10 Aα,10 bβ)]-, (-)-Salvinorin A, Divinorin A, Salvinorin, Salvinorin A |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | COC(=O)[C@@H]1 C[C@H](OC(=O)C)C(=O)[C@H]2 [C@@]1(C)CC[C@H]3 C(=O)O[C@@H](C[C@]23 C)c4cocc4 |
| IUPAC Name | methyl (2 S,4 aR,6 aR,7 R,9 S,10 AS,10 bR)-9-acetyloxy-2-(furan-3-yl)-6 A,10 b-dimethyl-4,10-dioxo-2,4 A,5,6,7,8,9,10 A-octahydro-1 H-benzo[f]isochromene-7-carboxylate |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction