missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac Xanthoanthrafil-d3, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | rac Xanthoanthrafil-d3 |
| CAS | 1216710-83-6 |
| Molecular Formula | C19H20D3N3O6 |
| Quantity | 10 mg |
| Synonym | N-[(3,4-Dimethoxyphenyl)methyl]-2-[(2-hydroxy-1-methylethyl)amino]-5-nitro-benzamide-d3, Benzamidenafil-d3 |
| InChI Formula | InChI=1S/C19H23N3O6/c1-12(11-23)21-16-6-5-14(22(25)26)9-15(16)19(24)20-10-13-4-7-17(27-2)18(8-13)28-3/h4-9,12,21,23H,10-11H2,1-3H3,(H,20,24)/i1D3 |
| SMILES | [2H]C([2H])([2H])C(CO)Nc1ccc(cc1C(=O)NCc2ccc(OC)c(OC)c2)[N+](=O)[O-] |
| IUPAC Name | N-[(3,4-dimethoxyphenyl)methyl]-5-nitro-2-[(1,1,1-trideuterio-3-hydroxypropan-2-yl)amino]benzamide |
| Molecular Weight (g/mol) | 392.42 |
| Formula Weight | 392.1775 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction