missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac Biotin-d4, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | (±)-Biotin-D4 (pentanoic acid 2,2,3,3-D4) |
| CAS | 1217850-77-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 D4 H12 N2 O3 S |
| Quantity | 10 mg |
| Synonym | rac Biotin-d4, (±)-Biotin-d4, rel-(3aR,4R,6aS)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanoic-2,2,3,3-d4 acid, 5-[(3aRS,4RS,6aSR)-2-Oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-2,2,3,3-tetradeuteriopentanoic acid, (±)-Biotin-D4 (pentanoic acid 2,2,3,3-D4) |
| InChI Formula | InChI=1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1/i1D2,2D2 |
| SMILES | [2H]C([2H])(C[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)C([2H])([2H])CC(=O)O |
| IUPAC Name | 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-3,3,4,4-tetradeuterio-pentanoic acid |
| Molecular Weight (g/mol) | 248.34 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction