missing translation for 'onlineSavingsMsg'
Learn More

(R)-Ketorolac, TRC

Product Code. 30008338
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008338 10 mg 10mg
30133741 25 mg 25mg
30137482 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008338 Supplier Toronto Research Chemicals Supplier No. TRCK23560010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66635-93-6
Molecular Formula C15H13NO3
Molecular Weight (g/mol) 255.27
Synonym 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, (1R)-, 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, (R)-, (1R)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid, (+)-Ketorolac, (R)-Ketorolac, R-(+)-Ketorolac
IUPAC Name (1R)-5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid
SMILES OC(=O)[C@@H]1CCn2c1ccc2C(=O)c3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H13NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13/h1-7,11H,8-9H2,(H,18,19)/t11-/m1/s1
Formula Weight 255.0895
Chemical Name or Material (R)-Ketorolac
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.