missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pregabalin, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Pregabalin
Specifications
Specifications
| Chemical Name or Material | Pregabalin |
| CAS | 148553-50-8 |
| Analyte or Component Names | Pregabalin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H17 N O2 |
| Synonym | Hexanoic acid, 3-(aminomethyl)-5-methyl-, (3 S)-, Hexanoic acid, 3-(aminomethyl)-5-methyl-, (S)-, (2 S)-3-(Aminomethyl)-5-methylhexanoic acid, (3 S)-3-(Aminomethyl)-5-methylhexanoic acid, 3 S-(aminomethyl)-5-methyl-hexanoic acid, (S)-(+)-3-(Aminomethyl)-5-methylhexanoic acid, (S)-3-(Ammoniomethyl)-5-methylhexanoate, (S)-Pregabalin, CI 1008, Gabanext 75, Lyrica, Mahagaba-75, Maxgalin 75, Neugaba 75, PD 144723, Pregabalin, Pregalin 75, (S)-Isobutylgaba |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CC(C)C[C@H](CN)CC(=O)O |
| IUPAC Name | (3 S)-3-(aminomethyl)-5-methylhexanoic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction