missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PFI 7
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PFI 7 is a selective GID4 antagonist (Kd = 80 nM by SPR assays and EC50 = 0.6 μM by cell-based NanoBRET). PFI 7 binds within the ?-barrel of GID4 substrate binding pocket, disrupting its interaction with the canonical Pro/N-degron peptide in live cells. It alters protein levels of several proteins including RNA helicases as measured by quantitative proteomics.
Specifications
Specifications
| Chemical Name or Material | N-((1s,4s)-4-(1H-Benzo[d]imidazol-2-yl)cyclohexyl)-2-(((1H-indol-2-yl)methyl)amino)acetamide dihydrochloride |
| Quantity | 10 mg |
| Target | Ubiquitin Ligase (E3) Inhibitors |
| Molecular Formula | C24H27N5O.2HCl |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction