missing translation for 'onlineSavingsMsg'
Learn More

Oxcarbazepine, Lipomed

Product Code. 30460236
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30460236

Brand: Lipomed LPMOXC175110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Oxcarbazepine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxcarbazepine
CAS 28721-07-5
Analyte or Component Names Oxcarbazepine
Shipping Condition Room Temperature
Molecular Formula C15 H12 N2 O2
Synonym Oxcarbazepine, 10,11-Dihydro-10-oxo-5 H-dibenz[b,f]azepine-5-carboxamide, GP 47680, Oxacarbazepine, Oxcarbazepine, Oxecarb, Oxetol, Trileptal
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES NC(=O)N1c2ccccc2CC(=O)c3ccccc13
IUPAC Name 5-oxo-6 H-benzo[b][1 ]benzazepine-11-carboxamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 252.27
Quantity 10 mg
Formula Weight 252.0899 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.