missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenimide, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Fenimide
Specifications
Specifications
| Chemical Name or Material | Fenimide |
| CAS | 60-45-7 |
| Analyte or Component Names | Fenimide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H15 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Ethyl-3-methyl-3-phenylpyrrolidine-2,5-dione, 2,5-Pyrrolidinedione, 4-ethyl-3-methyl-3-phenyl-, Succinimide, 3-ethyl-2-methyl-2-phenyl- (7 CI,8 CI), 3-Ethyl-2-methyl-2-phenylsuccinimide, 4-Ethyl-3-methyl-3-phenyl-2,5-pyrrolidinedione, C.I. 419, Fenimid, Fenimide, NSC 172121, NSC 248080, PM 1807, α-Methyl-α-phenyl-β-ethylsuccinimide |
| InChI Formula | InChI=1 S/C13H15NO2/c1-3-10-11(15)14-12(16)13(10,2)9-7-5-4-6-8-9/h4-8,10 H,3H2,1-2H3,(H,14,15,16) |
| SMILES | CCC1C(=O)NC(=O)C1(C)c2ccccc2 |
| IUPAC Name | 4-ethyl-3-methyl-3-phenylpyrrolidine-2,5-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction