missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrorphan, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Dextrorphan
Specifications
Specifications
| Chemical Name or Material | Dextrorphan |
| CAS | 125-73-5 |
| Analyte or Component Names | Dextrorphan |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H23 N O |
| Synonym | Morphinan-3-ol, 17-methyl-, (9α,13α,14α)-, 9α,13α,14α-Morphinan-3-ol, 17-methyl- (8 CI), (9α,13α,14α)-17-Methylmorphinan-3-ol, (+)-3-Hydroxy-N-methylmorphinan, (+)-Dextrorphan, (+)-17-Methyl-9 alpha,13 alpha,14 alpha-morphinan-3-ol, (+)-17-Methyl-9 alpha,13 alpha,14 alpha-morphinan-3-ol, (+)-17-Methyl-9 alpha,13 alpha,14 alpha-morphinan-3-ol, (+)-17-Methyl-9 alpha,13 alpha,14 alpha-morphinan-3-ol, (+)-17-Methyl-9 alpha,13 alpha,14 alpha-morphinan-3-ol, (+)-17-Methyl-9 alpha,13 alpha,14 alpha-morphinan-3-ol, (+)-17-Methyl-9 alpha,13 alpha,14 alpha-morphinan-3-ol |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CN1CC[C@@]23 CCCC[C@@H]2 [C@@H]1 Cc4ccc(O)cc34 |
| IUPAC Name | (1 S,9 S,10 S)-17-methyl-17-azatetracyclo[7.5.3.0 ^{1,10}0.0 ^{2,7}]heptadeca-2(7),3,5-trien-4-ol |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction