missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Erlotinib hydrochloride, MedChemExpress

Product Code. 16405598
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16405598

Brand: Medchem Express HY13256

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Desmethyl Erlotinib hydrochloride (OSI-420) is an active metabolite of Erlotinib. Erlotinib is a potent EGFR tyrosin kinase inhibitor.

Desmethyl Erlotinib hydrochloride (OSI-420) is an active metabolite of Erlotinib. Erlotinib is a potent EGFR tyrosin kinase inhibitor.

Specifications

Chemical Name or Material Desmethyl Erlotinib hydrochloride
CAS 183320-51-6
Color White
Purity Grade Notes Research
Molecular Formula C21H22ClN3O4
Quantity 10 mg
Synonym OSI-420CP-373420 hydrochloride
Solubility Information DMSO : 50 mg/mL (120.23 mM; Need ultrasonic)
SMILES OCCOC1=CC2=C(N=CN=C2NC3=CC=CC(C#C)=C3)C=C1OCCOC.Cl
Molecular Weight (g/mol) 415.87
Formula Weight 415.87
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.