missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Xylose-1,2,3,4,5,5'-C-d6, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | D-Xylose-1,2,3,4,5,5'-C-d6 |
| Molecular Formula | C5H4D6O5 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3+,4-,5?/m1/s1/i1D2,2D,3D,4D,5D |
| SMILES | OC1([2H])[C@@](O)([2H])[C@](O)([2H])[C@@](O)([2H])C([2H])([2H])O1 |
| Molecular Weight (g/mol) | 156.17 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction