missing translation for 'onlineSavingsMsg'
Learn More

AZM475271, MedChemExpress

Product Code. 16415758
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16415758

Brand: Medchem Express HY13561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZM475271 is a potent and selective Src kinase inhibitor with IC50 of 5 nM; no inhibitory activity on Flt3, KDR, Tie-2.

AZM475271 is a potent and selective Src kinase inhibitor with IC50 of 5 nM

Specifications

Chemical Name or Material AZM475271
CAS 476159-98-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H27ClN4O3
Quantity 10 mg
Synonym M475271
Solubility Information DMSO : ≥ 42 mg/mL (94.82 mM)
SMILES CN1CCC(COC2=CC3=NC=NC(NC4=CC(OC)=CC=C4Cl)=C3C=C2OC)CC1
Molecular Weight (g/mol) 442.94
Formula Weight 442.94
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.