missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Aprotinin
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
Description
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes.
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. The binding is reversible, and most aprotinin-protease complexes dissociate at a pH>10 or <3.2. Target enzumes include trypsin, chymatrypsin, kalikrein and plasmin. Activity: >4500 KIU/mg
Specifications
Specifica
| CAS | 9087-70-1 |
| Molecular Formula | C284H432N84O79S7 |
| Quantity | 10 mg |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto