missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amisulpride, Lipomed
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
missing translation for 'unitSize'
10mg
Description
Amisulpride
Specifications
Specifications
| Chemical Name or Material | Amisulpride |
| CAS | 71675-85-9 |
| Analyte or Component Names | Amisulpride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H27 N3 O4 S |
| Synonym | Amisulpride, 4-Amino-N-[[(2 RS)-1-ethylpyrrolidin-2-yl]methyl]-5-(ethylsulfonyl)-2-methoxybenzamide |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCN1CCCC1CNC(=O)c2cc(c(N)cc2OC)S(=O)(=O)CC |
| IUPAC Name | 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-ethylsulfonyl-2-methoxybenzamide |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction